missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, 0.25mm dia., Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10288BU

 View more versions of this product

Catalog No. AA10288BU



Specifications

Specifications

Platinum
7440-06-4
Pt
black
[Pt]
Odorless
23939
Wire, 0.25mm (0.01 in.) dia.
25cm
Silvery white wire
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
ISA Type R or S Standard

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents