missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, 0.1mm (0.004in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010959G5

 View more versions of this product

Catalog No. AA10959G5



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White
7440-06-4
21.45g/cm3 at 20°C
Pt
5 m
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Odorless
Platinum
Silvery white wire
99.997% (Metals basis)
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Wire

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only