missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, 0.1mm (0.004 in.) dia., 99.997% (metals basis), Alfa Aesar, Premion™

Manufacturer:  Thermo Fisher Scientific 10959G1

 View more versions of this product

Catalog No. AA10959G1



Specifications

Specifications

Platinum
7440-06-4
21.45g/cm3 at 20°C
Pt
black
[Pt]
Odorless
23939
Wire, 0.5mm (0.02 in.) dia.
1m
Silvery white wire
99.997% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents