missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, 99.95% (metals basis), Alfa Aesar™

Form: approx. 16.8g/m; Temper: as drawn

Manufacturer:  Alfa Aesar 10285BU

 View more versions of this product

Catalog No. AA10285BU



Specifications

Specifications

Platinum
Hard
Silvery white wire
99.95% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White
25cm
7440-06-4
21.45g/cm3 at 20°C
Pt
black
[Pt]
Odorless
23939
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents