missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.404mm dia., annealed, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar™ 45058CB

 View more versions of this product

Catalog No. AA45058CB



Specifications

Specifications

Platinum
7440-06-4
99.9% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
[Pt]
195.08
CHEBI:33400
250cm
Silvery white wire
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
Odorless
platinum
23939
Wire, 0.404mm (0.0159 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

SDS Documents