missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.404mm dia., annealed, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar™ 45058BW

 View more versions of this product

Catalog No. AA45058BW



Specifications

Specifications

Platinum
7440-06-4
21.45g/cm3 at 20°C
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
Odorless
platinum
23939
Wire, 0.404mm (0.0159 in.) dia.
50cm
Silvery white wire
99.9% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
[Pt]
195.08
CHEBI:33400

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

SDS Documents