missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 2.0mm (0.08 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10284BO

 View more versions of this product

Catalog No. AA10284BO



Specifications

Specifications

Platinum wire
1cm
7440-06-4
Pt
black
[Pt]
Odorless
23939
Wire
≈67.4g/m
2.0mm (0.0 8 in.) dia., Annealed, Temper: soft
(metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
99.95%

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents