missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Wire, 1.5mm (0.059 in.) dia., hard, 99.95% (Metals basis), Alfa Aesar™

Form: Approx.37.9g/m, Temper: as drawn

Manufacturer:  Alfa Aesar 13153BP

 View more versions of this product

Catalog No. AA13153BP



Specifications

Specifications

Platinum
Hard
Silvery white wire
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Wire, 1.5mm (0.06 in.) dia.
2cm
7440-06-4
99.95% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Documents