missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Wire, 1.0mm (0.04 in.) dia., Premion™, 99.997% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10956BP

 View more versions of this product

Catalog No. AA10956BP



Specifications

Specifications

Platinum
7440-06-4
99.997% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White
2cm
Silvery white wire
Pt
black
[Pt]
Odorless
23939
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Documents