Please login to your online account to display your discounted pricing

Platinum wire, 0.5mm (0.02in) dia, Premion™, 99.997% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Platinum wire
CAS 7440-06-4
Name Note 0.5mm (0.02 in.) dia, Premion™
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA10957BW

  SDS
alfa aesar
10957BW
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50cm
Chemical Name or Material Platinum wire
TSCA TSCA
Density ≈4.21g/m
Name Note 0.5mm (0.02 in.) dia, Premion™
Assay 99.997%
Assay Percent Notes (metals basis)
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless