Please login to your online account to display your discounted pricing

Platinum wire, 0.25mm dia., Alfa Aesar™

$361.88 - $1,573.54

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Purity Grade For ISA Type R or S Standard Grade
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10288BY

  SDS
alfa aesar
10288BY
100cm Each for $361.88

AA10288CC

  SDS
alfa aesar
10288CC
500cm Each for $1,573.54
Description & Specifications

Specifications

Quantity 100cm
Density 21.45g/cm3 at 20°C
Physical Form Wire, 0.25mm (0.01 in.) dia.
EINECS Number 231-116-1
Purity Grade For ISA Type R or S Standard Grade
Chemical Name or Material Platinum
MDL Number MFCD00011179
Appearance Silvery white wire
Odor Odorless
CAS 7440-06-4