missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Wire, 0.25mm (0.010 in.) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx.1.05g/m

Manufacturer:  Alfa Aesar 45093BY

 View more versions of this product

Catalog No. AA45093BY



Specifications

Specifications

Platinum
7440-06-4
99.9% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
[Pt]
195.08
CHEBI:33400
White
100cm
Silvery white wire
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
Odorless
platinum
23939
Wire, 0.25mm (0.01 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
SDS Documents