Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.127mm (0.005 in.) dia., For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple

Chemical Identifiers

Chemical Name or Material Platinum wire
CAS 7440-06-4
Name Note 0.127mm (0.005 in.) dia.
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10289G1

 
alfa aesar™
10289G1
1m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10289G5

 
alfa aesar™
10289G5
5m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10289G9

 
alfa aesar™
10289G9
25m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum wire
TSCA TSCA
Name Note 0.127mm (0.005 in.) dia.
Physical Form Wire
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless
Weight ≈0.27g/m
Quantity 1m