missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.127mm (0.005 in.) dia., For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple

Manufacturer:  Alfa Aesar™ 10289G9

 View more versions of this product

Catalog No. AA10289G9



Specifications

Specifications

Platinum wire
25m
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
≈0.27g/m
0.127mm (0.005 in.) dia.
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Wire

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents