missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Wire, 0.076mm (0.003 in.) dia., Alfa Aesar™

Form: Approx .0.045g/50cm

Manufacturer:  Thermo Fisher Scientific 10079G2

 View more versions of this product

Catalog No. AA10079G2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
30 wt% Rhodium wire
Silver colored wire
MFCD00198186
Wire, 0.076mm (0.003 in.) dia.
2m
7440-6-6
Pt:Rh; 70:30 wt%
Odorless
Silver

Chemical Identifiers

7440-6-6
Pt:Rh; 70:30 wt%
MFCD00198186
Documents