Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.0762mm (0.003 in.) dia., For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple

Chemical Identifiers

Chemical Name or Material Platinum wire
CAS 7440-06-4
Name Note 0.0762mm (0.003 in.) dia.
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10291BW

  SDS
alfa aesar™
10291BW
50cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10291G2

  SDS
alfa aesar™
10291G2
2m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10291G6

  SDS
alfa aesar™
10291G6
10m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum wire
TSCA TSCA
Name Note 0.0762mm (0.003 in.) dia.
Physical Form Wire
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless
Weight ≈0.05g/50cm
Quantity 50cm