missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum wire, 0.0762mm (0.003 in.) dia., For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple

Manufacturer:  Alfa Aesar™ 10291G6

 View more versions of this product

Catalog No. AA10291G6



Specifications

Specifications

Platinum wire
10m
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
≈0.05g/50cm
0.0762mm (0.003 in.) dia.
Pt
black
[Pt]
Odorless
23939
Wire

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents