missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Platinum wire, 0.0508mm (0.002 in.) dia., hard, 99.95% (metals basis)
Manufacturer: Alfa Aesar™ 13479G1
Specifications
Platinum wire | |
1m | |
7440-06-4 | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
Odorless | |
23939 | |
Wire |
≈0.043g/m | |
0.0508mm (0.002 in.) dia., Hard, Temper: as drawn | |
(metals basis) | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
195.08 | |
CHEBI:33400 | |
99.95% |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
TSCA : TSCA