missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Wire, 0.0508mm (0.002 in.) dia., 99.99% (Metals basis), Alfa Aesar™

Form: Approx.0.043g/m, Temper: soft

Manufacturer:  Alfa Aesar 43772G1

 View more versions of this product

Catalog No. AA43772G1



Specifications

Specifications

Platinum
7440-06-4
99.99% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White
1m
Silvery white wire
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Wire, 0.0508mm (0.002 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Documents