missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Tube, I.D x O.D: 1.32 x 1.57mm, , Alfa Aesar™

Form: Approx.0.62g/5cm

Manufacturer:  Thermo Fisher Scientific 10294BU

 View more versions of this product

Catalog No. AA10294BU



Specifications

Specifications

Platinum
7440-06-4
Pt
black
[Pt]
Odorless
23939
Tube, I.D X O.D: 1.32 X 1.57mm
25cm
Silvery white tube
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Documents