missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum Thinfoil, 0.004+/-0.0025mm (0.00016+/-0.0001 in.) thick, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 12868FF

 View more versions of this product

Catalog No. AA12868FF



Specifications

Specifications

Platinum Thinfoil
25 x 25mm
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Foil
≈0.05g/25x25mm
0.004 (±)0.0025mm (0.00016 (±)0.0001 in.) thick, Not certified pinhole free
(metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
99.95%

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents