Applied Biosystems™ TaqMan™ SARS-CoV-2 Plus Control
The Applied Biosystems TaqMan SARS-CoV-2 Plus Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 ORF1a, ORF1b, and N gene RUO assays.
Manufacturer: Applied Biosystems™ 956129
Description
The Applied Biosystems TaqMan SARS-CoV-2 Plus Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 ORF1a, ORF1b, and N gene RUO assays included in the:
- TaqMan SARS-CoV-2 with RNase P Assay 2.0 (Cat. No. A51331)
- TaqMan SARS-CoV2 MS2 RT-PCR Assay 2.0 (Cat. No. A51327)
- TaqMan SARS-CoV2 Fast PCR Assay 2.0 (Cat. No. A51246)
The control should be diluted in TaqMan SARS-CoV-2 Control Dilution Buffer (Cat. No. A49889).
Specifications
-20°C | |
TaqManTM SARS-CoV-2 with Rse P Assay 2.0 TaqManTM SARS-CoV-2 MS2 RT-PCR Assay 2.0 TaqMan™ SARS-CoV2 Fast PCR Assay 2.0 | |
TaqMan | |
Dry Ice |
Tube | |
1000 | |
10 x 10 μL |
For Research Use Only. Not for use in diagnostic procedures.