Learn More
Invitrogen™ Platinum™ SuperFi II PCR Master Mix
Invitrogen Platinum SuperFi II PCR Master Mix (2X) is a ready-to-use mixture of Platinum SuperFi II DNA Polymerase, Platinum SuperFi II Buffer, and dNTPs for convenient PCR setup and efficient amplification.
$461.00 - $9390.00
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | Platinum SuperFi II DNA Polymerase |
Reaction Speed | Fast |
Product Type | PCR Master Mix |
Shipping Condition | Dry Ice |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
12368010
|
Invitrogen™
12368010 |
100 Reactions |
Each for $461.00
|
|
|||||
12368050
|
Invitrogen™
12368050 |
500 Reactions |
Each for $2,090.00
|
|
|||||
12368250
|
Invitrogen™
12368250 |
5 x 500 Reactions |
Pack of 5 Each for $9,390.00
|
|
|||||
Description
Invitrogen Platinum SuperFi II PCR Master Mix (2X) is a ready-to-use mixture of Platinum SuperFi II DNA Polymerase, Platinum SuperFi II Buffer, and dNTPs for convenient PCR setup and efficient amplification. Platinum SuperFi II DNA Polymerase (Cat. No. 12361010) is a proofreading DNA polymerase that combines superior fidelity with innovative SuperFi II Buffer, enabling universal primer annealing for the highest success in PCR. It is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.
Features of Platinum SuperFi II PCR Master Mix include:
• Exceptional >300X Taq fidelity
• Universal primer annealing at 60°C
• Superior specificity, sensitivity, and yields
• Robust amplification of difficult-to-amplify targets (e.g., those with suboptimal purity, Ë65% GC content, long PCR requirement)
Platinum SuperFi II DNA Polymerase is an engineered enzyme with high processivity and increased resistance to PCR inhibitors. It also enables fast-cycling protocols and amplification of long targets (up to 20 kb). Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation. This technology also enables reaction setup at room temperature and provides increased sensitivity and yield.
Due to the unique composition of the SuperFi II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize the annealing temperature for each primer pair. With Platinum SuperFi II DNA Polymerase, different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.
Applications of Platinum SuperFi II DNA Polymerase:
• High-fidelity PCR
• Cloning and sub-cloning
• Site-directed mutagenesis
• Amplification of GC-rich templates
• Template generation for sequencing
• High-throughput PCR
• Amplification of samples with suboptimal purity
• Long PCR
• Fast PCR
Platinum SuperFi II Green PCR Master Mix (12369010) is also available, which additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels, further streamlining the PCR workflow from setup to final analysis.
Specifications
High | |
Fast | |
Dry Ice | |
2X | |
Built-In Hot Start | |
SuperMix or Master Mix |
Platinum SuperFi II DNA Polymerase | |
PCR Master Mix | |
Hot-start PCR, High-fidelity PCR | |
>300X | |
Blunt | |
20 kb or less |