Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Premion™, 99.99+% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum slug
CAS 7440-06-4
Name Note 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
Purity Grade Premion™
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13374KG

  SDS
alfa aesar™
13374KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum slug
TSCA TSCA
Name Note 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
Purity Grade Premion™
Physical Form Slug
Assay ≥99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless
Weight ≈8.61g/slug
Quantity 1 piece