Please login to your online account to display your discounted pricing

Platinum Ruthenium black, nominally Pt 50%, Ru 50% (Atomic wt%), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Platinum Ruthenium black
Name Note HiSPEC 6000
Physical Form Solid
Molecular Formula Pt:Ru; 50:50
MDL Number MFCD00798552
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4117103

  SDS
alfa aesar
4117103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4117106

  SDS
alfa aesar
4117106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41171MD

  SDS
alfa aesar
41171MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Color Black
DOT Information DOT Class: 4.1, Packaging Group: II
Odor Odorless
MDL Number MFCD00798552
Chemical Name or Material Platinum Ruthenium black
Physical Form Solid
Health Hazard 2 Keep container tightly closed and dry. Keep away from sources of ignition - No Smoking. Take precautionary measures against static discharges.
Health Hazard 1 H228: Flammable solid.
Name Note HiSPEC 6000
Molecular Formula Pt:Ru; 50:50
Safety and Handling

H228: Flammable solid.

Keep container tightly closed and dry. Keep away from sources of ignition - No Smoking. Take precautionary measures against static discharges.