Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum rod, 6.35mm (0.25 in.) dia., Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum rod
CAS 7440-06-4
Name Note 6.35mm (0.25 in.) dia.
Purity Grade Premion™
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12556HF

  SDS
alfa aesar™
12556HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum rod
TSCA TSCA
Name Note 6.35mm (0.25 in.) dia.
Purity Grade Premion™
Physical Form Rod
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless
Weight ≈6.8g/cm
Quantity 10mm