missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 12216FF

 View more versions of this product

Catalog No. AA12216FF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pt:Rh:Pd; 90:5:5 wt%
≈0.39g/25x25mm
Woven from 0.076mm (0.003 in.) dia. wire
MFCD00011179
99.9%
Platinum Rhodium Palladium gauze
25 x 25mm
(metals basis)
80 Mesh
Documents