missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)
Manufacturer: Alfa Aesar™ 12216GH
Specifications
Pt:Rh:Pd; 90:5:5 wt% | |
≈0.39g/25x25mm | |
Woven from 0.076mm (0.003 in.) dia. wire | |
MFCD00011179 | |
99.9% |
Platinum Rhodium Palladium gauze | |
100 x 100mm | |
(metals basis) | |
80 Mesh |