missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 12217FF

 View more versions of this product

Catalog No. AA12217FF



Specifications

Specifications

Pt:Rh; 90:10 wt%
≈0.39g/25x25mm
Woven from 0.076mm (0.003 in.) dia. wire
MFCD00198186
Odorless
Platinum Rhodium gauze
25 x 25mm
(metals basis)
80 Mesh
99.9%
Documents