Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum Rhodium gauze
Name Note Woven from 0.076mm (0.003 in.) dia. wire
MDL Number MFCD00198186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12217FF

 
alfa aesar™
12217FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12217FI

 
alfa aesar™
12217FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12217GH

 
alfa aesar™
12217GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum Rhodium gauze
TSCA TSCA
Name Note Woven from 0.076mm (0.003 in.) dia. wire
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00198186
Mesh Size 80 Mesh
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Weight ≈0.39g/25x25mm
Quantity 25x25mm