missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003 in.) dia. wire, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 12217FI

 View more versions of this product

Catalog No. AA12217FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pt:Rh; 90:10 wt%
≈0.39g/25x25mm
Woven from 0.076mm (0.003 in.) dia. wire
MFCD00198186
Odorless
Platinum Rhodium gauze
50 x 50mm
(metals basis)
80 Mesh
99.9%
Documents