Learn More
Platinum powder, amorphous, APS <3 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum powder, 99.9%, Pt, CAS Number-7440-06-4, platinum, metal, black, metal, platino, platine, iv ion, sponge, platinum, elemental, platin german, platin, 5g, (metals basis), CHEBI:33400, Hazard Class: 4.1; Packaging Group: III, 231-116-1, H228, P210-P240-P241-P280-P370+P378q
Supplier: Thermo Scientific Chemicals 03670403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum powder | |
Amorphous Powder | |
(metals basis) | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
99.9% |
7440-06-4 | |
1 g | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Odorless |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-116-1
RTECSNumber : TP2160000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only