missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum powder, amorphous, APS 2.0-8.0 micron, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 47145MC

 View more versions of this product

Catalog No. AA47145MC



Specifications

Specifications

Platinum
100mg
(metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
[Pt]
195.08
CHEBI:33400
Amorphous Powder
0.5 to 1.5m2/g
Tap density: 0.5 to 2.5g/cm3
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
Odorless
platinum
23939
APS 2.0 to 8.0 micron
99.9%

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

RTECSNumber : TP2160000

TSCA : TSCA

SDS Documents