Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum powder, amorphous, <10 micron, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum powder
CAS 7440-06-4
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1210003

  SDS
alfa aesar™
1210003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1210006

  SDS
alfa aesar™
1210006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1210077

  SDS
alfa aesar™
1210077
0.1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Amorphous Powder
Odor Odorless
Particle Size <10 Micron
Quantity 1g
MDL Number MFCD00011179
RTECS Number TP2160000
TSCA TSCA
Assay 99.9%
Assay Percent Notes (metals basis)
Weight Tap density: 0.9g/cm3 to 1.5g/cm3
Chemical Name or Material Platinum powder
CAS 7440-06-4
Surface Area 9.2 to 15.7m2/g