missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum powder, amorphous, <10 micron, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1210006

 View more versions of this product

Catalog No. AA1210006



Specifications

Specifications

Platinum powder
(metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Amorphous Powder
Odorless
9.2 to 15.7m2/g
Tap density: 0.9g/cm3 to 1.5g/cm3
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
99.9%
<10 Micron
5g

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

RTECSNumber : TP2160000

TSCA : TSCA

Documents