Learn More
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Material For Gas Sensors
Supplier: Thermo Scientific Chemicals 04544506
Description
Platinum, 5% on activated carbon paste is used for a broad range of reactions common to palladium on carbon catalysts. it is utilized for hydrogenolysis reactions using molecular hydrogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum | |
7440-06-4 | |
Pt | |
5 g | |
Insoluble in water. | |
[Pt] | |
195.08 | |
CHEBI:33400 |
5% on graphite | |
Powder | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
Powder |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-116-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only