missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on alumina powder, reduced, Alfa Aesar™

Manufacturer:  Alfa Aesar™ 4422206

 View more versions of this product

Catalog No. AA4422206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
5% on alumina powder
Powder
Powder
5g
7440-6-6
MFCD00011179

Chemical Identifiers

7440-6-6
MFCD00011179
Safety and Handling

Safety and Handling

H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS Documents