missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum, 5% on graphite, Type 286, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4436514

 View more versions of this product

Catalog No. AA4436514



Specifications

Specifications

Platinum
5% on graphite
Powder
MFCD00011179
Soluble in water.
[Pt]
195.08
CHEBI:33400
25g
7440-06-4
Pt
black
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Powder

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H228-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents