missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1067201

 View more versions of this product

Catalog No. AA1067201



Specifications

Specifications

Platinum powder
(metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Powder
Premion™
Odorless
0.25g
7440-06-4
Pt
black
[Pt]
195.08
CHEBI:33400
99.99%
−22 Mesh
Silver-White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

RTECSNumber : TP2160000

TSCA : TSCA

Documents