missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum powder, -22 mesh, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01067203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum powder | |
Silver-White | |
1 g | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Premion™ |
7440-06-4 | |
Powder | |
(metals basis) | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
99.99% | |
Odorless |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-116-1
RTECSNumber : TP2160000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only