missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum powder, -200+400 mesh, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 4683003

 View more versions of this product

Catalog No. AA4683003



Specifications

Specifications

Platinum powder
(metals basis)
MFCD0001117
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Powder
−200+400 Mesh
1g
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
99.98%
Odorless

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD0001117
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

RTECSNumber : TP2160000

TSCA : TSCA

SDS Documents