missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Potassium Chloride (Laboratory), Fisher Chemical

Manufacturer:  Thermo Scientific P1585

 View more versions of this product

Catalog No. P158-5




Specifications

Specifications

Platinum Potassium Chloride
Cl6K2Pt
3288
DPAIVKJGTXERIM-UHFFFAOYSA-H
dipotassium;hexachloroplatinum(2-)
61856
Powder/Solid
3.499g/cm³
Glass Bottle
250°C
16921-30-5
MFCD00011389
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]
485.981
486.01
Laboratory
Pass Test
Yellow to Orange
5g

Chemical Identifiers

16921-30-5
485.981
DPAIVKJGTXERIM-UHFFFAOYSA-H
61856
Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]
Cl6K2Pt
MFCD00011389
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
dipotassium;hexachloroplatinum(2-)
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

Documents