missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14397AE

 View more versions of this product

Catalog No. AA14397AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
Liquid
MFCD00011179
Characteristic
100mL
7440-6-6
Pt in 20% HCl
Liquid
Approx. 1g/cm3

Chemical Identifiers

7440-6-6
Pt in 20% HCl
MFCD00011179
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

SDS Documents