missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13827AP

 View more versions of this product

Catalog No. AA13827AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
Clear liquid
MFCD00011179
Liquid
Colorless
7440-6-6,7647-01-0,7732-18-5
Pt in 20% HCl
Hydrochloric acid: 20.0%; Antimony: 0.1%; Water: 79.9%
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

7440-6-6
Pt in 20% HCl
MFCD00011179
Safety and Handling

Safety and Handling

H314-H319-H335: Causes severe skin burns and eye damage. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-595-7

Documents