missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, nominally 70% on high surface area advanced carbon support, HiSPEC™ 13100

Manufacturer:  Alfa Aesar™ 4504103

 View more versions of this product

Catalog No. AA4504103



Specifications

Specifications

Platinum, HiSPEC™ 13100
Gray to Black
Nominally 70% on high surface area advanced carbon support
Pt
UN1325
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
Odorless
1g
7440-06-4
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
195.08
CHEBI:33400

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
Pt
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
CHEBI:33400
Safety and Handling

Safety and Handling

GHS H Statement
H228-H351
Flammable solid.
Suspected of causing cancer.

GHS P Statement
P210-P280-P281-P308+P313-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents