missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, nominally 60% on high surface area advanced carbon support, HiSPEC™ 9100

Manufacturer:  Alfa Aesar™ 4472806

 View more versions of this product

Catalog No. AA4472806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum, HiSPEC™ 9100
5g
7440-06-4
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
195.08
CHEBI:33400
Odorless
Nominally 60% on high surface area advanced carbon support
Pt
UN1325
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
Pt
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
CHEBI:33400
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents