missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support, HiSPEC™ 12100

Manufacturer:  Alfa Aesar™ 4504003

 View more versions of this product

Catalog No. AA4504003



Specifications

Specifications

Platinum
1g
7440-6-6
UN1325
Gray to Black
Nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support, HiSPEC™ 12100
MFCD00798552

Chemical Identifiers

7440-6-6
MFCD00798552
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents