missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, nominally 40%, Ruthenium, nominally 20% on carbon black, HiSPEC™ 10000

Manufacturer:  Alfa Aesar™ 4417209

 View more versions of this product

Catalog No. AA4417209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
10g
7440-6-6
UN1325
Black
Nominally 40%, Ruthenium, nominally 20% on carbon black, HiSPEC™ 10000
MFCD00798552
Powder

Chemical Identifiers

7440-6-6
MFCD00798552
Safety and Handling

Safety and Handling

GHS H Statement
H228-H351
Flammable solid.
Suspected of causing cancer.

GHS P Statement
P210-P280-P281-P308+P313-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents