Please login to your online account to display your discounted pricing

Platinum, nominally 40% on carbon black, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Platinum
Assay Percent Range nominally 40% on carbon black
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4220403

  SDS
alfa aesar
4220403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4220406

  SDS
alfa aesar
4220406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Platinum
MDL Number MFCD00011179
Assay Percent Range nominally 40% on carbon black