missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum(IV) sulfide, Pt 74.8% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01399206
Description
Ultra high purity and high purity compositions of platinum sulfide improve both optical quality and usefulness as scientific standards.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(IV) sulfide | |
12038-21-0 | |
5 g | |
MFCD00016293 | |
Insoluble in water. | |
S=[Pt]=S | |
259.20 | |
259.22 |
225°C to 250°C (decomposition) | |
Powder | |
PtS2 | |
platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt | |
RKNFJIIYAUSTJA-UHFFFAOYSA-N | |
bis(sulfanylidene)platinum | |
82862 | |
Pt 74.8% min. |
Chemical Identifiers
12038-21-0 | |
259.20 | |
RKNFJIIYAUSTJA-UHFFFAOYSA-N | |
82862 | |
S=[Pt]=S |
PtS2 | |
MFCD00016293 | |
platinum iv sulfide, platinum sulfide, disulfanylideneplatinum, bis sulfanylidene platinum, dithioxoplatinum, platinum disulphide, acmc-20ajvg, platinum iv sulfide 1g, platinic sulfide,, platinum iv sulfide, pt | |
bis(sulfanylidene)platinum |
Safety and Handling
EINECSNumber : 234-876-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only